Specifications
- Wavelength selection:Automatic, based on method selection
- Description:DR6000 Benchtop Spectrophotometer without RFID Technology
- Enclosure rating:IP20 with closed lid
- Storage conditions:−25...+60 °C / max. 80% relative humidity (non-condensing)
- Sample cell compatability:Rectangular: 10, 20, 30, 50 mm, 1 inch; round: 13 mm, 16 mm, 1 inch
- User programs:200
- Wavelength accuracy:±1 nm
- Spectral bandwidth:2 nm
- Wavelength reproducibility:<0.1 nm
- Optical design:Reference beam, spectral
- Stray light:KI-solution at 220 nm <3.3 Abs/ <0.05%
- Photometric accuracy:5 mAbs @ 0.0 - 0.5 Abs
<1% @ 0.5 - 2.0 Abs @ 546 nm - Weight:11 kg
- Width:500 mm
- Display:TFT 7" WVGA color touch
- Depth:460 mm
- Wavelength range:190 - 1100 nm
- Wavelength resolution:0.1 nm
- Operating mode:Transmittance (%), absorbance and concentration (wavelength, time)
- Photometric linearity:0.005 - 2 Abs
≤0.01 at > 2 Abs with neutral glass at 546 nm - Photometric measuring range:± 3 Abs
- Datalog capacity (sets):5000 data points (result, date, time, sample-ID, user-ID)
- Height:215 mm
- Lamp source, lifetime:Tungsten (visible range), deuterium (UV range)
- Languages:En, es, fr, ja, ko, pt, zh (in manual); Bg, cn, cz, da, en, es, fr, gr, hr, hu, it, jp, kr, nl, pl, pt, ro, ru, sl, sl, sv, tr (User interface languages)
- Method storage:>240 (Preprogrammed)
- Operating conditions:10 - 40 °C, max. 80% relative humidity (non-condensing)
- Power requirements:100 - 240 V; 50/60 Hz
- Scan speed:900 nm/min (in 1 nm steps)
- Cat. no.:CA11027-712
- Electrical:100/240 V, 50/60 Hz
- Supplier No.:LPV44199.00002
Specifications
About this item
The DR6000™ is the industry's most advanced lab spectrophotometer. It offers high-speed wavelength scanning across the UV and Visible Spectrum, and comes with over 250 pre-programmed methods including the most common testing methods used today. With optional accessories allowing for high-volume testing via a carousel sample changer, and increased accuracy with a sample delivery system that eliminates optical difference errors, this instrument ensures you are ready to handle your wide-ranging water testing needs.
- Optional RFID Technology, for easy method update, sample ID, and certificate of analysis
- Your water testing needs, all in one spectrophotometer
- Accessories for high volume and high accuracy testing needs
- Advanced quality assurance at your fingertips
- Guided procedures and elimination of false readings
- Automatically avoids errors
Your Water Testing Needs, All in One Spectrophotometer—The DR6000 has the most pre-programmed testing methods, including high-speed wavelength scanning across the UV and Visible Spectrum.
Accessories for High Volume and High Accuracy Testing Needs—A carousel sample changer allows up to seven sequential measurements. The Sipper Module, an instrument-controlled sample delivery system, increases precision by constant optical characteristics.
Advanced Quality Assurance at Your Fingertips—The DR6000 comes with integrated QA software for scheduling, documenting and interpreting all of your needed quality measurements.
Guided Procedures and Elimination of False Readings—The DR6000, when used with TNTplus® reagent vials, provides the accurate results you need by guiding you step-by-step through your testing procedures. With TNTplus, the instrument averages 10 readings and eliminates outliers, making scratched, flawed or dirty glassware a non-issue.
Note: RFID technology currently available only in US, Anguilla, American Samoa, Australia, Bolivia, Canada, Cayman Islands, Columbia, Dominican Republic, El Salvador, Federated States of Micronesia, Guam, Guatemala, Marshall Islands, New Zealand, Northern Mariana Islands, Palau, Panama, Puerto Rico, and US Virgin Islands.