Mix and Go!™ Competent Cells, Zymo Research
Fournisseur: Zymo Research
Certificats
A propos de cet article
The Mix and Go!™ Competent Cells are pre made, chemically competent cells for simple and highly efficient DNA transformation.
- Mix and Go!™ transformation procedure with transformation efficiencies of 108 - 109 transformants/μg of plasmid DNA
- Simply add DNA and then spread
- DNA transformation in as little as 20 seconds
- Uses: bacterial transformations, DNA cloning, blue-white screening
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.