96-Well PCR Conversion Plates, Zymo Research
Fournisseur: Zymo Research
Certificats
A propos de cet article
Conversion plates for use with EZ-96 DNA Methylation™ bisulfite conversion kits.
- 96-well, non-skirted PCR plates with easy-to-read alphanumeric labels
- Rimmed wells minimize cross contamination
- Provided with or without adhesive, pierceable foil cover
Customers Who Bought This Also Bought