Anti-IgG Lama Polyclonal Antibody (Horseradish Peroxidase (HRP))
# de catalogue 75783-570
Fournisseur: Immunoreagents
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Secondary
- Clonality:Polyclonal
- Gel shift:Yes
- Host:Llama
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:IgG
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:1.0 mg
- Western blot:Yes
- Format:Purified IgG
- Form:Lyophilized
- Storage buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2-8 °C
- Concentration:1.0 mg/ml (E 1% at 280 nm = 13.0)
- Shipping temperature:2-8 °C
- Immunogen:Mouse IgG
- Purification:Affinity purified using solid phaseMouse IgG
- Cat. no.:75783-570
- Supplier No.:IR2980
Spécifications
A propos de cet article
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Horseradish Peroxidase (HRP)
Epitope:
Host: Llama
Isotype: IgG
Reactivity: Mouse
Customers Who Bought This Also Bought