SiliaSep™ Tosic Acid (SCX) Flash Cartridges, SiliCycle
Fournisseur: SiliCycle
Certificats
A propos de cet article
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
- Highest silica gel quality
- Innovative packing technology
- Universal Luer fittings
- Compatible with any flash system
- Cost effective
- Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from milligrams to kilograms.
Customers Who Bought This Also Bought