AimTab™ Bluing Tablets, Germaine® Laboratories
# de catalogue 75910-964
Fournisseur: Germaine Laboratories
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:AimTab™ Bluing Tablets
- Cat. no.:75910-964
- Supplier No.:40021
Spécifications
A propos de cet article
Tablets are designed for use in all drug testing collections to prevent adulteration
- Aids in detecting adulteration of drug tets
- Meets federal DOT regulations 49 CFR 40.41
- Helps detect leaks in plumbimg systems
Can also be used by plumbers to detect leaks. Not classified as an In-Vitro Diagnostic test.
Customers Who Bought This Also Bought