ContactUsLinkComponent
Gemini 88 Plus Dual Rate Syringe Pump, KD Scientific
Gemini 88 Plus Dual Rate Syringe Pump, KD Scientific
# de catalogue 76000-914
Fournisseur:  KD Scientific
Gemini 88 Plus Dual Rate Syringe Pump
Gemini 88 Plus Dual Rate Syringe Pump, KD Scientific
# de catalogue 76000-914
Fournisseur:  KD Scientific
Spécifications
  • Description:
    Gemini 88 Plus Dual Rate Syringe Pump
  • Cat. no.:
    76000-914
Spécifications
A propos de cet article
The Gemini 88 Plus Dual Rate Syringe Pump is really two syringe pumps in one by providing two independent pumping channels linked through hardware and an intuitive 7” touch screen interface. This dual rate syringe pump can infuse simultaneously at different rates, or infuse with one syringe and withdraw with the other. When combined with a valve box, it provides the continuous delivery of a peristaltic pump with the accuracy and low flow rates of a syringe pump.
  • Two separate syringe pumps in one instrument
  • Large touch screen display allows quick and easy setup
  • High or low pressure operation
  • Compatible with a wide variety of syringes
  • Continuous fluid delivery
  • TTL, USB and RS-232 communication
The intuitive Gemini 88 Plus graphical user interface controlled with a large 7" LCD color touch screen display allows quick and easy setup
Three operating conditions named Independent, Reciprocating and Twin are available as standard features to satisfy various applications in life science and industrial settings. Independent Condition allows the Gemini 88 Plus to operate as two separate syringe pumps named P1 and P2. Each syringe will operate independently with different syringe types, sizes, force, or target (volume or time, mode dependent). In Reciprocating Condition, both syringe channels move in opposite directions at the same rate using the same syringe size and type. When combined with a valve box, the Reciprocating Condition can provide the continuous fluidic delivery or a peristaltic pump with the accurate, pulseless, low flow rates provided by a syringe pump. Twin Condition allows both syringes to operate in the same mode using the exact same syringe type, syringe size, force, target (volume or time) and flow rate settings. The pump also allows the user to combine both flows for higher speed and volume infusion applications.
The display run screen presents the user with all key dispensing parameters in real time. Syringe tables containing all major syringe manufacturers allow simple selection of any compatible syringe size. Audible Alarms, Adjustable Force and Screen Lock are all features that are available with a touch of the screen.
Certifications: CE, ETL (UL and CSA), CB Scheme, EU RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.
Customers Who Bought This Also Bought