A propos de cet article
FreeZone Small Tray Dryers provide precise control and program flexibility for consistent performance and are designed for lyophilizing small to moderate bulk sample loads or numerous sample batches.
- Compact design for small bulk loads or numerous sample batches
- Precision with 3 heated shelves up to +60°C
- Flexibility: temperature and vacuum level controls in 30 programs with up to 16 segments each
- Quality reassurance with onboard data collection and real time graphing
- Sample probes with remote notification of operating parameters or alerts
Three 10.6” x 11.7” heated shelves with sensors for monitoring sample temperature provide 372 sq
Lyo-Works™ OS provides flexible programming and control with a real time display of freeze dryer and tray dryer parameters on a 5"" full color touch screen on the connected FreeZone Freeze Dryer (sold separately). One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.
Samples must be pre-frozen.
inches of shelf space with the option for 2 additional shelves (sold separately). Each shelf warms to +60°C and a microprocessor maintains the system temperature within 3°C of set point. A clear acrylic viewing door provides complete visibility of the shelves.
Spécifications
- Catalog No:
- 76000-446
- 76000-448
- 76000-450
- 76000-438
- 76000-440
- 76000-442
- 76000-444
- Environmentally Preferable:
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
- Depth:
- 470 mm (18.5")
- 470 mm (18.5")
- 470 mm (18.5")
- 470 mm (18.5")
- 470 mm (18.5")
- 470 mm (18.5")
- 470 mm (18.5")
- Height:
- 458.5 mm (18.05")
- 458.5 mm (18.05")
- 458.5 mm (18.05")
- 458.5 mm (18.05")
- 458.5 mm (18.05")
- 458.5 mm (18.05")
- 458.5 mm (18.05")
- Width:
- 388.6 mm (15.3")
- 388.6 mm (15.3")
- 388.6 mm (15.3")
- 388.6 mm (15.3")
- 388.6 mm (15.3")
- 388.6 mm (15.3")
- 388.6 mm (15.3")