Thermal Microbeads, Electron Microscopy Sciences
# de catalogue 103302-222
Fournisseur: Electron Microscopy Sciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Thermal Microbeads, 1 L, 2.5 - 3 mm
- Cat. no.:103302-222
- Supplier No.:68605-10
Spécifications
A propos de cet article
Effectively heat and cool samples in most water and dry baths.
- Reduce sample contamination in applications requiring water and ice
- Eliminates the need for floating racks and bottle weights
- Autoclavable and solvent resistant
- For use in water/drybaths, ice pans/ buckets, microbead chambers and microbead bag
Use of the microbeads reduces sample contamination in applications requiring water and ice and provides support for tubes and bottles.
Customers Who Bought This Also Bought