A propos de cet article
Heated product shelves with clear chamber for drying of bulk samples or small containers.
- 10" diameter shelves are removable
- Shelf temperature may be set from -20 °C (-4 °F) to 60 °C (140 °F)
- Shelves may be placed in freezer for pre-freezing
- Shelves are height adjustable
For use with any FreeZone Freeze Dryer except Legacy models
Each Chamber includes a 12" diameter clear acrylic cylinder, vacuum release valve and 8" diameter opening to collector on bottom of chamber. Will fit 10.12" or 14.0" diameter condensers. Short Chambers include three shelves and three temperature probes. Tall Chambers include five shelves and five temperature probes.
Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates. The 10" diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing. Each shelf is height adjustable to accommodate a broad variety of sample sizes. Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature from the Lyo-Works display.
Spécifications
- Catalog No:
- 76000-946
- 76000-948
- 76000-950
- 76000-952
- 76000-958
- 76000-940
- 76000-942
- 76000-944
- 76000-932
- 76000-934
- 76000-978
- 76000-936
- 76000-954
- 76000-956
- Environmentally Preferable:
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials