ContactUsLinkComponent
FreeZone® 18 L Console Freeze Dryers, Labconco®
FreeZone® 18 L Console Freeze Dryers, Labconco®
Fournisseur:  Labconco
   Sustainable Options Available
Certificats
FreeZone® 18 L Console Freeze Dryers, Labconco®
FreeZone® 18 L Console Freeze Dryers, Labconco®
Fournisseur:  Labconco
   Sustainable Options Available
Certificats
Spécifications
  • Catalog No:
  • 76000-188
  • 76000-144
  • 76000-186
  • 76000-142
  • 76000-184
  • 76000-140
  • 76000-182
  • 76000-138
  • 76000-152
  • 76000-150
  • 76000-192
  • 76000-148
  • 76000-190
  • 76000-146
  • 76000-180
  • 76000-562
  • 76000-564
  • 76000-178
  • 76000-176
  • 76000-566
  • 76000-154
  • 76000-166
  • 76000-164
  • 76000-162
  • 76000-160
  • 76000-174
  • 76000-172
  • 76000-170
  • 76000-168
  • 76000-552
  • 76000-554
  • 76000-158
  • 76000-156
  • 76000-556
  • 76000-558
  • 76000-560
  • Environmentally Preferable:
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health
  • Temperature:
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • -50°C (-58°F), Collector
  • Depth:
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • 751.8 mm (29.6")
  • Height:
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • 929.6 mm (36.6")
  • Width:
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")
  • 807.7 mm (31.8")