Avantor® Apollo Silica, HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations at an economical price.
- Easy scale-up from analytical to prep
- Suitable for pH 2 to 8
Columns are ideal for routine analyses and for educational laboratories.
Specifications
- Catalog No:
- 76398-198
- 75797-726
- 75830-362
- USP code:
- L3
- L3
- L3