Avantor® Apollo Phenyl, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations.
- Easy scale-up from analytical to preparative
- Economical price
Columns are ideal for routine analyses and for educational laboratories.
Spécifications
- Catalog No:
- 75797-720
- 76398-202
- 75797-718
- 75797-722
- 75830-366
- 75797-716
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11