Avantor® Hichrom, Organic Acid HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Hichrom organic acid columns are ion exclusion columns packed with sulphonated polystyrene-divinylbenzene.
- Rapid analysis of organic acids and alcohols
- pH stable polymer resin
- Isocratic 100% aquoeus mobile phases only, no organic solvents
- USP L17
The OA-1000 and OA-2000 columns exhibit excellent selectivity for aliphatic and aromatic acids.
The IOA-1000 and IOA-2000 columns are suitable for the separation of citric and other acids from glucose and fructose.
As with most ion exclusion columns, a column heater is necessary for normal operating procedures.
Customers Who Bought This Also Bought