Wire Nuts
# de catalogue 470092-054
Fournisseur: MOUSER ELECTRONICS SE
Spécifications
- Voltage:300 V
- Description:Gray Wire Nuts, Female, insulated with Polyvinyl Chloride
- Packaging:Bulk
- Length:8.51 mm
- Supplier No.:ME159-2216