Big Trap Gas Purifier, Trajan Scientific and Medical
Fournisseur: Trajan Scientific and Medical
Certificats
A propos de cet article
Big Trap Gas Purifiers are deisgned for bulk purification applications, or where several instruments are plumbed from a single source.
- One-piece, heavy walled aluminium tube
- 750 cm³ capacity
- Pressure rating up to 250 psig
Customers Who Bought This Also Bought