Gas Purifier Kit, Trajan Scientific Americas
Fournisseur: Trajan Scientific and Medical
Certificats
A propos de cet article
Gas purifier kits are designed to provide fast stabilization times to reduce gas consumption and provide clean gas to GC and GCMS systems.
- Gas purifiers and mounting unit in a complete kit
- Full removal of gas impurities for different applications
- Improved trace analysis
Customers Who Bought This Also Bought