Gas Purifier, Trajan Scientific Americas
Fournisseur: Trajan Scientific and Medical
Certificats
A propos de cet article
Gas purifiers are designed to provide fast stabilization times to reduce gas consumption and provide clean gas to GC and GCMS systems.
- Removal of oxygen, moisture and hydrocarbons that can impact analytical results
- Fast stabilization times
- Improved trace analysis
Customers Who Bought This Also Bought