ContactUsLinkComponent
SP Wilmad-LabGlass PEEK Union, SP Industries
SP Wilmad-LabGlass PEEK Union, SP Industries
# de catalogue 14007-326
Fournisseur:  SP Industries
UNION PEEK 1/4-28
SP Wilmad-LabGlass PEEK Union, SP Industries
# de catalogue 14007-326
Fournisseur:  SP Industries
Spécifications
  • Description:
    PEEK Union
  • Cat. no.:
    14007-326
  • Supplier No.:
    BP-1824-018
Spécifications
A propos de cet article
This PEEK (polyether ether ketone) union is used for connecting 1/4"-28 fittings.
Ordering information: Nuts and ferrules included.
Customers Who Bought This Also Bought