Digital Temperature Control, Time Proportional Two-Outlet, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Compact, time-proportional digital temperature controller with two 120vac front outlets
- Controller for use with Type J temperature probes with a male SMP/OST plug
- Ideal for two-circuit heating mantles
- Love control module and CAT5 patch cable available for PC control and data logging
Digital temperature controller sold as controller only, or as a complete item with a probe and lead. Controller has two front outlets that provide 1800 watts total from either plug, or in combination, and one rear alarm outlet. Features include and over-temp cutoff, RFI free power, and 0.1 degree C accuracy. 120vac 50/60Hz input.
Also sold in this group are a USB signal converter and a CAT5 patch cable for use with the controller.
Ordering information: One year limited warranty.
Customers Who Bought This Also Bought