Scale-Up Series Bench-Top Reactor System, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Ace Glass Scale-Up Series plain bench-top reactor system includes the essential items needed to begin building a reactor for your specific application.
- Flask made of high-quality borosilicate glass.
- Reactors are sized for bench-top use, and their geometry translates into ACE Kilo Scale Reactors.
- Base system includes a flask, a CAPFE o-ring, and a stand with supports.
Scale-Up Series reactors are available in 100 mL to 6000 mL capacities, and due to their geometry, the reaction can be translated into Kilo Scale reactors.
Ordering information: Base systems include the essential items, and can be customized by adding a head, condenser, funnel, heating mantle, or other items, which are sold separately.
Customers Who Bought This Also Bought