Avantor® Allsep, Anion IC HPLC Columns, 7 µm
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Allsep Anion is a methacrylate based phase with quaternary ammonium functional groups, optimized for use with both suppressed and non-suppressed conductivity detection.
- 7 µm polymer based anion exchange phase
- Suppressed or non-suppressed conductivity detection
- pH range 2 to 10
- Use with 0 to 100% organic modifier
- USP L23
Columns are compatible with common IC mobile phases, such as carbonate, bicarbonate, p-hydroxybenzoic acid, phthalic acid, succinic acid and sodium octane sulfonate.
Allsep Anion is recommended for applications involving inorganic anions, weak and strong acid ions, metal complexes and organic acids.
Certifications: Meets the requirements for the EPA method 300.0 Part A for determining inorganic ions in water.
Spécifications
- Catalog No:
- 76398-332
- 75797-894
- 76398-210
- 75798-962
- 76398-212
- 76398-208
- 75797-888
- 75830-604
- 75797-892
- 75797-890
- pH range:
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- USP code:
- L23
- L23
- L23
- L23
- L23
- L23