Accessories for TLC Developing Chamber, General Glass Blowing
Fournisseur: General Glass Blowing
Certificats
Certificats
A propos de cet article
Customers Who Bought This Also Bought
Methanol ≥99.8%, GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®
Fournisseur: MilliporeSigma
$578.24 to $2,015.51
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$146.85 to $576.51
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Fournisseur: VWR International
$147.27 to $464.44
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$377.44 to $992.90
Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Ricca Chemical Company
Fournisseur: Ricca Chemical
$162.71