Avantor® Hichrom, Anion Exclusion HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Hichrom Anion Exclusion columns are used for the separation of organic acids and weakly ionised anions.
- Polymer-based for broad pH stability
- USP L22
- Stainless steel construction
Mobile phases contain dilute mineral acids. Small amounts of acetonitrile are added to decrease retention of hydrophobic molecules.
Customers Who Bought This Also Bought