J-KEM® Dual Temperature Controllers, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
J-Kem® Scientific Apollo dual temperature controller features two independent temperature controllers in a single cabinet.
- Temperature ramping and serial communications are also provided with this unit
Each independent channel has its own high power outlet, 100 hour timer to turn heating/cooling on or off at a user set time, and solid state over temperature protection circuit. .
Ordering information: Types T,J and K are supplied complete with PTFE Thermocouple (1/8 x 12"), Coiled Extension Cord (10') and 14/20 PTFE Thermocouple Adapter. The RTD Unit is the controller only.
Spécifications
- Catalog No:
- 89166-646
- 89166-644
- 89166-648
- Electrical:
- 10 Amps @ 120VAC, 1200 Watts MAX/Channel, 1800 Watts (Total)
- 10 Amps @ 120VAC, 1200 Watts MAX/Channel, 1800 Watts (Total)
- 10 Amps @ 120VAC, 1200 Watts MAX/Channel, 1800 Watts (Total)
- W×D×H:
- 8×9×4"
- 8×9×4"
- 8×9×4"