Support Stand for CHEMRxnHUB™ Bench Top Reactors, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
This unique stationary reactor stand is designed to fit into most benchtop hoods
Stand can hold a single reactor. The stand provides support for the reaction vessel(s), glassware, overhead stirring motor, and temperature monitor. Stand includes removable spill tray.
Customers Who Bought This Also Bought