Viscometer Cleaning and Drying Apparatus, Kohler
Fournisseur: Koehler
Certificats
A propos de cet article
It cleans and dries glass capillary viscometers using solvent and pressurized filtered air.
- Six tube capacity
- For all types of capillary viscometers
Adjustable drainage rack is used to drain excess sample oil from tubes prior to cleaning. Drainage trough connects to a suitable waste container or chemical drain for removal of waste oil and solvent. Built-in air filter removes particles from the air stream. Available (optional) solvent tank has tubing with fittings for connection to apparatus. Requires pressurized air source (150 psi maximum).
Customers Who Bought This Also Bought