ContactUsLinkComponent
Neutrophil Elastase Activity Assay Kit, Cayman Chemical Company
Neutrophil Elastase Activity Assay Kit, Cayman Chemical Company
# de catalogue 75817-036
Numéro CAS:  
Neutrophil Elastase Activity Assay Kit
Neutrophil Elastase Activity Assay Kit, Cayman Chemical Company
# de catalogue 75817-036
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Kit contains: Buffers, Reagents, and 96-Well Plate
  • Cat. no.:
    75817-036
  • Supplier No.:
    600610-192
Spécifications
A propos de cet article
A convenient all-in-one tool to measure neutrophil elastase activity.
  • Neutrophils isolated from whole blood (protocol included)
  • Measure neutrophil elastase released by neutrophils after various experimental conditions
  • Employs the specific elastase substrate (Z-Ala-Ala-Ala-Ala)2Rh110
  • Includes reagents to isolate neutrophils from whole blood and PMA to stimulate elastase release
  • Assay performed in provided black 96-well plates
Neutrophil elastase is stored within cytoplasmic azurophilic granules in the neutrophil and released upon stimulation by pathogens where it acts either as free protein or is associated with networks of extracellular traps (NET). Together with other proteases released from activated neutrophils, neutrophil elastase plays a critical role in degrading invading pathogens and thus provides the earliest line of defense in the immune system. It is thought to play a critical role in tumor invasion and metastasis as well as other inflammatory conditions. Cayman’s Neutrophil Elastase Activity Assay kit is designed to be used to study compounds regulating elastase release in neutrophils. The kit employs a specific non-fluorescent elastase substrate, (Z-Ala-Ala-Ala-Ala)2Rh110, which is selectively cleaved by elastase to yield the highly fluorescent compound R110, which can be analyzed with an excitation wavelength of 485 nm and emission wavelength of 525 nm. Reagents needed to isolate neutrophils from whole blood are included in the kit, as is PMA, which is known to stimulate elastase release from neutrophils.
BodyTextAccessories: Neutrophil (mouse) Isolation Kit (Item No. 601070) available to isolate mouse neutrophils from peritoneal lavage or bone marrow.
Customers Who Bought This Also Bought