Glas-Col® Heating Mantle, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Fabric heating mantle fits reaction flasks ranging from 100mL to 6000mL, including Ace Scale-Up Series flasks.
- Fabric exterior saves space
- Fabric interior reduces the chance of thermal shock
- Withstands 400 degrees C internal operating temperature
Certifications: CSA Approved.
Customers Who Bought This Also Bought