HiVac Valves, 3-Way PTFE Tip, Chemglass
# de catalogue 89094-266
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Valve with 3-Way PTFE Tip
- Cat. no.:89094-266
- Bore Size:0 - 4 mm PTFE stopcock
- Supplier No.:CGK-557-01
Spécifications
A propos de cet article
High vacuum valve features a three-way configuration for use in the construction of manifolds and adapters.
Customers Who Bought This Also Bought