DuoClick™ Specimen Container, Azer Scientific
Fournisseur: Azer Scientific Inc.
Certificats
A propos de cet article
DuoClick™ containers made of 100% virgin grade polypropylene ensure safe transport of specimen.
- Graduated
- Sterile tab label
- Seal is fully engaged with an audible 'click'
- Ideal for for pneumatic tube transport
- Visual cues on the container and lid aid in correct closure of container
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.