Wide-Neck Jerricans with/without Threaded Connector, Burkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Wide-neck canisters feature an extremely wide, 88 mm (3.46") filling opening.
- Carrying handles and srew lid guarantee safe and convenient transport
- Wide-neck jerrican can also be used as a settling canister
- Transparent HDPE construction
- UV stabilized
Models with threaded connector feature a threaded connector just above the base, where a spigot or other accessories with an inner thread of ³/₄" can be attached.
Jerricans are easy to clean on the inside, which is particularly important when storing sensitive substances.
Customers Who Bought This Also Bought