Forced Air Ovens, SM05, SHEL LAB
Fournisseur: Sheldon Manufacturing
Certificats
A propos de cet article
The SMO5 ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning and life cycle testing.
- Embedded microprocessor control with countdown timer
- Horizontal forced airflow
- 1.75" (4.5 cm) access port
- 24 month limited warranty
The oven is equipped with a stainless steel interior for long life operation and easy cleaning
This unit has a quick ramp-up time to 306°C for faster cycle times. This unit includes an advanced PID temperature control system for sensitive response.
With triple wall construction, this oven’s walls minimize external wall temperature which meets CE requirements to keep work-spaces cool.
Certifications: DIN 12880 Compliant.
Customers Who Bought This Also Bought