Magid® Gemstone Diamond Protective Visitor Glasses, Magid Glove
Fournisseur: Magid Glove
Certificats
A propos de cet article
These glasses feature a contemporary wrap-around frame with a clear polycarbonate lens.
- Visitor spec
- Vented sideshields
- UV protective glasses fit comfortably over prescription glasses
The Y20C spectacle also utilizes an impact-resistant lens. Glasses provide 99.9% UV protection.
Certifications: Meet ANSI Z87.1-2010 High Impact Standards.
Packaging: Each pair is wrapped in a sealed polybag.
Customers Who Bought This Also Bought