Magid® KnitMaster® Half-Finger Machine Knit Nylon Gloves, Magid Glove
Fournisseur: Magid Glove
Certificats
A propos de cet article
These Magid's KnitMaster gloves made of 100% nylon, seamless machine knit glove perfect for general-purpose use.
- Half-finger style for greater dexterity
- Excellent in environments in which lint is a problem
- Color-coded overedge to prevent unraveling in wash
The KnitMaster series is great for paint departments and as liners for the electronic and computer industries.
Ideal where lint is a problem, this glove features a long-lasting, tight knit reversible style with a knit wrist that resists unraveling.
Spécifications
- Catalog No:
- 11000-444
- 11000-442
- 11000-440
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Gauge:
- 10
- 10
- 10