KIMAX® KimCote® Culture Tubes, Reusable, with Unattached Caps, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
KimCote® plastic-coated culture tubes are made from tubing with uniform wall thickness for maximum heat transfer and chemical resistance.
- Ideal for biohazard test protocols
- Supplied with phenolic screw-thread caps, unattached
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The safety coating reduces the hazards of shattered glass and leakage of toxic or hazardous materials.
Certifications: ASTM E982, Type VI, Class A requirements.
Frequently Bought Together