Two-Piece Pressure Reactor with Jacketed Flask, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Complete, two-piece pressure reactor system with jacketed flask, designed to perform low to moderate positive pressure reactions, synthesis and catalysis.
- Jacketed flask made of borosilicate glass
- Two-piece design allows the use of large agitators and facilitates product removal and clean-out
- Reactor uses Ace-Thred inlet/outlets to achieve a leak-tight system
- Rated 35psig at 100 degrees C
Complete pressure reactor system with two-piece design offers a flanged flask and head to allow for the use of large agitators, and to facilitate product removal and clean-out. The system includes a jacketed flask, a flanged multi-neck head, an addition funnel, a condenser, a pressure manifold, and a super magnet stirring assembly.
Ordering information: Stand and support shelf not included.
Customers Who Bought This Also Bought