Airgas® Manifold Protocol Station
Fournisseur: Airgas
Certificats
A propos de cet article
These two-station manifolds are designed to provide a safe, reliable method of wall-mounting a single cylinder regulator, eliminating the need to handle the regulator during cylinder change out.
- Made from 304 stainless steel
- Durable, long-life service
- Easily mount to wall or existing structure
Manifolds are provided with 36" stainless steel (corrugat-ed bellows) flexible pigtails and CGA connections with check valves to prevent contaminates from entering during cylinder change-out.
Customers Who Bought This Also Bought