Complete Scintillation Probe Assembly
# de catalogue 470220-454
Fournisseur: SPECTRUM TECHNIQUES, LLC
Spécifications
- Description:Complete Scintillation Probe Assembly
- Cat. no.:470220-454
- Supplier No.:SPA38
Spécifications
A propos de cet article
Gamma Spectroscopy detector.
- Unit provides a complete detector assembly for gamma spectroscopy applications
The 38x38 mm NaI(Tl) crystal includes PMT and voltage divider as an integral assembly. The probe is supplied complete with a 10-position sample stand, lead shield and cables for connecting directly to the spectrometer. Resolution is typically 7% @ 662 keV.
Ordering information: Includes scintillation detector, 38x38mm with voltage divider; GMS38 position stand with sample tray and LS038 lead shield; and BNC6 and MHV6 cable assemblies.
Customers Who Bought This Also Bought