Duran GL45 Cap Membrane for Pressure Applications, Ace Glass Incorporated
# de catalogue 75877-646
Fournisseur: Ace Glass
Spécifications
- Description:Pressure Cap Membrane for GL45 Connection System
- Cat. no.:75877-646
- Supplier No.:7632-08
Spécifications
A propos de cet article
Cap membrane for pressure use is part of the GL45 connection system used with Duran lab bottles.
Cap membrane filter for use in pressure equalization as part of the Duran GL45 Connection System. GL45 Connection Systems are used when a safe transfer of liquid media within a closed and sterile system is required.
Customers Who Bought This Also Bought