Spécifications
- Description:Benchtop Meter-Attached Electrode Stand
- Cat. no.:89206-426
- Supplier No.:STARA-BEA
Spécifications
A propos de cet article
This meter-attached electrode stand can be used to easily place sensors in samples.
- Simplifies rinsing and storage between samples
- Cable management system minimizes electrode breakage
- 360° motion allows electrodes to be moved vertically, horizontally, and front-to-back positioning into beakers or sample containers
Can be attached to either side of Orion™ Versa Star benchtop meters and Orion™ Star™ A210 and Star™ A110 series benchtop meters. Can also be used with weighted base for compatibility with any meter system (portable or benchtop).