VAI® MP Garments, Veltek
Fournisseur: Veltek Associates
Certificats
A propos de cet article
The MP line is constructed using a non-woven, microporous, coated material that provides even greater personal protection in the cleanroom.
- Highly breathable fabric releases heat while maintaining a high level of protection
- Garments feature elastic in high movement areas for ease of movement and appropriate fit
- 100% bound seam construction for increased durability and reduced particle permeability
- Individually packaged in easy to tear bags with two liner bags per case
- Packaging is labled with lot number and expiration date
All MP coveralls are equipped with elastic wrists and ankles, along with a zipper closure. All MP frocks feature elastic wrists and a zipper closure. The breathable fabric provides protection against low level splashes and sprays, offering reliable fluid and liquid resistance. All sterile garments are gamma irradiated at 10⁻⁶ SAL.
Ordering information: All garments are supplied with a one-year warranty.
Packaging: All garments are individually packaged, with the exception of the MP Knee-High boots, which are packaged in pairs.
Spécifications
- Catalog No:
- 76308-426
- 76308-422
- 76308-424
- 76308-418
- 76308-420
- 75795-408
- 75795-544
- 75795-636
- 75795-642
- 75795-644
- 75795-402
- 75795-646
- 75795-354
- 75795-404
- 75795-584
- Model:
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- 1700 Easy2Gown
- Garment application:
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory