Airguard® Synthetic Pocket Filters, Total Filtration
Fournisseur: Total Filtration Services
Certificats
A propos de cet article
Extended surface pocket filters feature efficiency, high dust holding capacity, long service life, and low resistance to air flow.
Applications include general ventilation, stand-alone single-stage filtration, built-up frame banks, side-access systems, rooftop air handlers, air supply for paint spray booths, and economical upgrade from fiberglass or synthetic-framed disposable filters.
Ordering information: Extended surface pocket filters are available in ratings from MERV 9 to MERV 14; contact your Avantor representative for more information.
Customers Who Bought This Also Bought