Universal Computer Spectrometer
# de catalogue 470220-452
Fournisseur: SPECTRUM TECHNIQUES, LLC
Spécifications
- Description:Universal Computer Spectrometer
- Cat. no.:470220-452
- Supplier No.:UCS30-1K
Spécifications
A propos de cet article
This analyzer connects to a USB port eliminating the need to install cards inside the computer.
- Reduces the need for a computer card
This unit aids in the identification of nuclides, it contains a unique peak labeling feature named ISOMATCH. Providing the unit is accurately energy calibrated, the user may select a nuclide from the library and the corresponding characteristic emission lines are superimposed on the spectrum along with isotope and energy information. Using this feature, users may quickly check a spectrum and visually identify the emission components for each nuclide present.
Customers Who Bought This Also Bought