Kilo Scale Thermocouple Probe Adapter, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Anti-whip, glass thermocouple probe adapter for protecting ¼" O.D
- Heavy borosilicate glass
- Available as complete unit or glass sheath only
- Complete item includes nylon bushing, U10 Ace-Thred tap, and one #7 nylon adapter
The heavy glass sheath is topped with a 45/50 standard taper joint with a #25 Ace-Thred. Complete unit also includes one #25 nylon bushing with a #7 Ace-Thred tap, and one #7 nylon adapter designed for use with ¼" O.D. probes.
probes within kilo scale reactors.
Customers Who Bought This Also Bought