Dairy Bottles, High Density Polyethylene, Environmental Express®
Fournisseur: Environmental Express
Certificats
A propos de cet article
Included caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE.
- Leakproof design keeps contents safe inside
- Familiar and easy pouring with the milk-type jug design
Customers Who Bought This Also Bought