Pigment Lance, Burkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Pigment Lances are designed to sample pigments, pastes, and agglomerating bulk goods.
- With half-open flute (50 %) for heavily sticking products
- With quarter-open flute (25 %) for lightly sticking products
- Stainless steel AISI 316L (1.4404)
- 30 mm diameter
Packaging: One lance per package.
Customers Who Bought This Also Bought