Bottles, Aluminum, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
The aluminum bottle is particularly suited for storing, transporting, or sending substances and reference samples which have to be stored completely unaltered
- Pure aluminum (material AL 99.5)
- With tamper-evident screw cap made of PP, with aluminum inner seal
- Suitable for food (exception: highly acidic and salt-containing foods)
- With UN approval (for 60 to 5600 ml [2.03 to 189.36 oz.])
These bottles can be completely emptied without residues. They are odor-neutral, impervious to light and rustproof.
Customers Who Bought This Also Bought