Spunbonded Polypropylene Shoe Covers, SCR200 Series, CT International
Fournisseur: CT International
Certificats
A propos de cet article
SCR200 Series spunbonded polypropylene shoe covers are manufactured from a lightweight material that offers breathability and comfort, while maintaining full coverage
- Strong and durable
- Contamination control
- No latex proteins
- Lot traceability
All items are individually inspected for stitching, sizing, material, quality, and workmanship.
Packaging: Stacked and rolled, 50 Pairs/Bag, 4 Bags/Case, 200 Pair/Case.
Spécifications
- Catalog No:
- 56613-804
- 33502-676
- 33502-674
- Fabric:
- Spun-Bonded Polypropylene
- Spun-Bonded Polypropylene
- Spun-Bonded Polypropylene
- Garment application:
- Laboratory
- Laboratory
- Laboratory