Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
These probe maintenance kits help ensure optimal performance of Orion™ polarographic dissolved oxygen probes.
Ordering information: Each probe maintenance kit includes two membrane caps, one electrolyte solution, and a polishing disk.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.